Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198850.1 | 3941 | UTR 3 | NP_001185779.1 | ||
NM_001198851.1 | 3941 | UTR 3 | NP_001185780.1 | ||
NM_001198852.1 | 3941 | UTR 3 | NP_001185781.1 | ||
NM_014947.4 | 3941 | UTR 3 | NP_055762.3 | ||
XM_005270632.4 | 3941 | UTR 3 | XP_005270689.1 | ||
XM_006710458.3 | 3941 | UTR 3 | XP_006710521.1 | ||
XM_006710459.3 | 3941 | UTR 3 | XP_006710522.1 | ||
XM_011541026.2 | 3941 | UTR 3 | XP_011539328.1 | ||
XM_017000693.1 | 3941 | UTR 3 | XP_016856182.1 | ||
XM_017000694.1 | 3941 | UTR 3 | XP_016856183.1 | ||
XM_017000695.1 | 3941 | UTR 3 | XP_016856184.1 |