Product Details

SNP ID
rs11552341
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:161072116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGGAGACAGGTTCCCTCACAC[T/C]GGAGGCAGATTCCAGCTCCAGCTAT
Phenotype
MIM: 614264 MIM: 609607
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030916.2 2273 UTR 3 NP_112178.2
XM_005245508.3 2273 UTR 3 XP_005245565.1
XM_011510021.2 2273 UTR 3 XP_011508323.1
XM_011510022.2 2273 UTR 3 XP_011508324.1
XM_011510023.1 2273 Intron XP_011508325.1

View Full Product Details