Product Details

SNP ID
rs11551255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19523929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACGCGCCTTGGCCGGGCCCGG[T/G]GCCACGCAGGTGGTGCACTGCGCGC
Phenotype
MIM: 603465 MIM: 602101
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
There are no transcripts associated with this gene.

Gene
CLDN5
Gene Name
claudin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130861.1 797 Silent Mutation GCA,GCC A194A NP_001124333.1
NM_003277.3 797 Silent Mutation GCA,GCC A194A NP_003268.2
XM_017028929.1 797 Silent Mutation GCA,GCC A194A XP_016884418.1
XM_017028930.1 797 Silent Mutation GCA,GCC A194A XP_016884419.1

View Full Product Details