Product Details

SNP ID
rs11568615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62980944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATTGTGTCTCCTCCTGCCCCA[A/G]CCAGCCTTTTGTCTCAGTCTGTCTG
Phenotype
MIM: 607582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC22A6 PubMed Links

Gene Details

Gene
SLC22A6
Gene Name
solute carrier family 22 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004790.4 Intron NP_004781.2
NM_153276.2 Intron NP_695008.1
NM_153277.2 Intron NP_695009.1
NM_153278.2 Intron NP_695010.1
XM_017018562.1 Intron XP_016874051.1

View Full Product Details