Product Details

SNP ID
rs11575108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6822656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGAAGAGTCAAGGTCTCCCCA[C/T]GGGGAGTCAGCACCTTATTGAACAT
Phenotype
MIM: 186940 MIM: 610342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD4 PubMed Links

Gene Details

Gene
CD4
Gene Name
CD4 molecule
There are no transcripts associated with this gene.

Gene
GPR162
Gene Name
G protein-coupled receptor 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014449.1 Intron NP_055264.1
NM_019858.1 Intron NP_062832.1
Gene
P3H3
Gene Name
prolyl 3-hydroxylase 3
There are no transcripts associated with this gene.

View Full Product Details