Product Details

SNP ID
rs11574158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:31033837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACTGTGTGCGCCGGGGAGGCGCC[A/G]GCTTGTACTCGGCAGCGCGGGAATA
Phenotype
MIM: 604611
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PURG PubMed Links

Gene Details

Gene
PURG
Gene Name
purine rich element binding protein G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015508.2 576 Intron NP_001015508.1
NM_001323311.1 576 Intron NP_001310240.1
NM_001323312.1 576 Intron NP_001310241.1
NM_013357.2 576 Intron NP_037489.1
Gene
WRN
Gene Name
Werner syndrome RecQ like helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000553.4 576 UTR 5 NP_000544.2
XM_011544639.2 576 UTR 5 XP_011542941.1
XM_011544640.1 576 Intron XP_011542942.1

View Full Product Details