Product Details

SNP ID
rs180690065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71106366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTAGGCAACTTTACTTTACTG[C/T]GGATGTAATACAGCAACACGACGAC
Phenotype
MIM: 300033 MIM: 308380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025265.2 318 Missense Mutation CAC,CGC H29R NP_001020436.1
XM_005262244.3 318 Missense Mutation CAC,CGC H29R XP_005262301.1
Gene
FOXO4
Gene Name
forkhead box O4
There are no transcripts associated with this gene.

Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
There are no transcripts associated with this gene.

View Full Product Details