Product Details

SNP ID
rs180698014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50275960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGGGTGTTCAAGGAGTCCGTGT[A/G]CGCCTGGGGGGTGACGGGACAGTCA
Phenotype
MIM: 602898 MIM: 604293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
There are no transcripts associated with this gene.

Gene
PLXNB2
Gene Name
plexin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012401.3 5434 Missense Mutation NP_036533.2
XM_005261909.1 5434 Missense Mutation XP_005261966.1
XM_005261910.2 5434 Missense Mutation XP_005261967.1
XM_005261911.4 5434 Missense Mutation XP_005261968.1
XM_006724413.2 5434 Missense Mutation XP_006724476.1
XM_011530682.2 5434 Missense Mutation XP_011528984.1
XM_017028703.1 5434 Missense Mutation XP_016884192.1
XM_017028704.1 5434 Missense Mutation XP_016884193.1

View Full Product Details