Product Details

SNP ID
rs180832748
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:117147749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGAGAAGCCTTTGAGTTTAGC[A/G]GCTGTAGGTGACTCCGCCTTTTGAT
Phenotype
MIM: 608162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VTCN1 PubMed Links

Gene Details

Gene
VTCN1
Gene Name
V-set domain containing T cell activation inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253849.1 1173 Intron NP_001240778.1
NM_001253850.1 1173 Intron NP_001240779.1
NM_024626.3 1173 Intron NP_078902.2
XM_011542143.2 1173 Missense Mutation CCG,CTG P303L XP_011540445.2
XM_017002335.1 1173 Missense Mutation CCG,CTG P258L XP_016857824.1

View Full Product Details