Product Details

SNP ID
rs181151514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127197262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCTCCCTCAGAATGGGCCTCC[A/G]GGTTCGGTATTTGGGCACCTTTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf56 PubMed Links

Gene Details

Gene
C3orf56
Gene Name
chromosome 3 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007534.2 817 Missense Mutation AGG,GGG R193G NP_001007535.1

View Full Product Details