Product Details

SNP ID
rs181167667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42103981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCGTGAGGCTCAAGCTCACCGT[C/G]CAGGACCCCGGGTTTCCCATGGGCA
Phenotype
MIM: 613859
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UMODL1 PubMed Links

Gene Details

Gene
UMODL1
Gene Name
uromodulin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004416.2 4337 Silent Mutation GTC,GTG V471V NP_001004416.2
NM_001199527.1 4337 Silent Mutation GTC,GTG V399V NP_001186456.1
NM_001199528.2 4337 Silent Mutation GTC,GTG V399V NP_001186457.2
NM_173568.3 4337 Silent Mutation GTC,GTG V471V NP_775839.3
XM_011529797.1 4337 Silent Mutation GTC,GTG V293V XP_011528099.1
XM_017028506.1 4337 Silent Mutation GTC,GTG V399V XP_016883995.1
XM_017028507.1 4337 Silent Mutation GTC,GTG V367V XP_016883996.1
XM_017028508.1 4337 Intron XP_016883997.1
Gene
UMODL1-AS1
Gene Name
UMODL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details