Product Details

SNP ID
rs180870479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132078015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCATGGGTGACCTTATTTCTT[C/T]GCCATACAAAGCATAGTAGTCTGGT
Phenotype
MIM: 300628 MIM: 300547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMD7 PubMed Links

Gene Details

Gene
FRMD7
Gene Name
FERM domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306193.1 2142 Missense Mutation AAA,GAA K653E NP_001293122.1
NM_194277.2 2142 Missense Mutation AAA,GAA K668E NP_919253.1
XM_017029947.1 2142 Missense Mutation AAA,GAA K652E XP_016885436.1
XM_017029948.1 2142 Missense Mutation AAA,GAA K583E XP_016885437.1
XM_017029949.1 2142 Missense Mutation AAA,GAA K510E XP_016885438.1
Gene
STK26
Gene Name
serine/threonine protein kinase 26
There are no transcripts associated with this gene.

View Full Product Details