Product Details

SNP ID
rs180848748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:7231728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTTGCACGTGGGCTCGAATC[G/T]CCTGGAAAACATCGCTGAGAGCATC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC30 PubMed Links

Gene Details

Gene
LRRC30
Gene Name
leucine rich repeat containing 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105581.1 590 Missense Mutation CGC,CTC R197L NP_001099051.1

View Full Product Details