Product Details

SNP ID
rs180994734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49881501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGTGGTTCAAGTGTGATGATGC[C/T]GTCATCACTAAGGCCAGTATTAAGG
Phenotype
MIM: 300975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP27X PubMed Links

Gene Details

Gene
USP27X
Gene Name
ubiquitin specific peptidase 27, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145073.2 1635 Silent Mutation GCC,GCT A398A NP_001138545.1
Gene
USP27X-AS1
Gene Name
USP27X antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details