Product Details

SNP ID
rs181000076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122789149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCCTGGCCAACCTCTCCCTG[A/C]TGGATGCCTGCTATTCCTCCGCCAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5C1 PubMed Links

Gene Details

Gene
OR5C1
Gene Name
olfactory receptor family 5 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001923.1 217 Missense Mutation ATG,CTG M73L NP_001001923.1

View Full Product Details