Product Details

SNP ID
rs181385267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180849107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATGATTGATAAGACTTGAGTG[A/T]GAGATGTATGCCTTCCCACACTCAT
Phenotype
MIM: 610281
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFP62 PubMed Links

Gene Details

Gene
ZFP62
Gene Name
ZFP62 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172638.1 2558 Silent Mutation TCA,TCT S796S NP_001166109.1
NM_152283.4 2558 Silent Mutation TCA,TCT S763S NP_689496.4
XM_017009710.1 2558 Silent Mutation TCA,TCT S763S XP_016865199.1
XM_017009711.1 2558 Silent Mutation TCA,TCT S763S XP_016865200.1
XM_017009712.1 2558 Silent Mutation TCA,TCT S763S XP_016865201.1
XM_017009713.1 2558 Silent Mutation TCA,TCT S763S XP_016865202.1
XM_017009714.1 2558 Silent Mutation TCA,TCT S763S XP_016865203.1
XM_017009715.1 2558 Silent Mutation TCA,TCT S763S XP_016865204.1
XM_017009716.1 2558 Intron XP_016865205.1
XM_017009717.1 2558 Silent Mutation TCA,TCT S796S XP_016865206.1
XM_017009718.1 2558 Intron XP_016865207.1
XM_017009719.1 2558 Intron XP_016865208.1
XM_017009720.1 2558 Intron XP_016865209.1
XM_017009721.1 2558 Intron XP_016865210.1

View Full Product Details