Product Details

SNP ID
rs181689044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123753855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTGAAGGAATTCGTAGGATTG[C/T]GGACAGAATGTAGATATAAGAAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6X1 PubMed Links

Gene Details

Gene
OR6X1
Gene Name
olfactory receptor family 6 subfamily X member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005188.1 664 Missense Mutation ACA,GCA T222A NP_001005188.1

View Full Product Details