Product Details

SNP ID
rs181483445
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:182399990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCATCAGCCAGGAGCCCATCA[G/T]CAGAATCAGAAAGGTCTCCATCAGC
Phenotype
MIM: 138290
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLUL PubMed Links

Gene Details

Gene
GLUL
Gene Name
glutamate-ammonia ligase
There are no transcripts associated with this gene.

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

Gene
TEDDM1
Gene Name
transmembrane epididymal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172000.3 627 Missense Mutation NP_741997.3

View Full Product Details