Product Details

SNP ID
rs181511686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101397598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAATATCTTCAGAGCTTTGGG[A/G]TACATCTGATATCTGAATCACAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREG2 PubMed Links

Gene Details

Gene
CREG2
Gene Name
cellular repressor of E1A stimulated genes 2
There are no transcripts associated with this gene.

Gene
RFX8
Gene Name
RFX family member 8, lacking RFX DNA binding domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145664.1 1501 Missense Mutation CCC,TCC P458S NP_001139136.1
XM_011511771.2 1501 Missense Mutation CCC,TCC P534S XP_011510073.1
XM_011511772.1 1501 Missense Mutation CCC,TCC P529S XP_011510074.1
XM_011511773.1 1501 Missense Mutation CCC,TCC P428S XP_011510075.1
XM_011511776.1 1501 Missense Mutation CCC,TCC P362S XP_011510078.1
XM_011511777.1 1501 Missense Mutation CCC,TCC P362S XP_011510079.1
XM_011511778.1 1501 Missense Mutation CCC,TCC P362S XP_011510080.1
XM_017004851.1 1501 Missense Mutation CCC,TCC P571S XP_016860340.1
XM_017004852.1 1501 Missense Mutation CCC,TCC P500S XP_016860341.1
XM_017004853.1 1501 Intron XP_016860342.1
XM_017004854.1 1501 Intron XP_016860343.1

View Full Product Details