Product Details

SNP ID
rs181544365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55755571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGCGTACTACCATATTTCTAT[A/G]TTTTTTCAAAATGACATTGGAGCTG
Phenotype
MIM: 112265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP5 PubMed Links

Gene Details

Gene
BMP5
Gene Name
bone morphogenetic protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249304.3 1224 UTR 3 XP_005249361.1
XM_011514816.2 1224 Missense Mutation CAT,TAT H406Y XP_011513118.1
XM_011514817.2 1224 Intron XP_011513119.1
XM_017011198.1 1224 Missense Mutation CAT,TAT H228Y XP_016866687.1

View Full Product Details