Product Details

SNP ID
rs181910006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49662015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGGGACCTCGCCATCAGGCCC[A/G]TGCCCGCTGTTGGGGAGCAGCTCCT
Phenotype
MIM: 610837 MIM: 603734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L12 PubMed Links

Gene Details

Gene
BCL2L12
Gene Name
BCL2 like 12
There are no transcripts associated with this gene.

Gene
IRF3
Gene Name
interferon regulatory factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197122.1 1226 Silent Mutation CAC,CAT H305H NP_001184051.1
NM_001197123.1 1226 Silent Mutation CAC,CAT H270H NP_001184052.1
NM_001197124.1 1226 Intron NP_001184053.1
NM_001197125.1 1226 Silent Mutation CAC,CAT H159H NP_001184054.1
NM_001197126.1 1226 Silent Mutation CAC,CAT H159H NP_001184055.1
NM_001197127.1 1226 Intron NP_001184056.1
NM_001197128.1 1226 Intron NP_001184057.1
NM_001571.5 1226 Silent Mutation CAC,CAT H305H NP_001562.1
XM_006723197.1 1226 Silent Mutation CAC,CAT H305H XP_006723260.1
XM_006723198.1 1226 Silent Mutation CAC,CAT H305H XP_006723261.1
XM_006723200.1 1226 Silent Mutation CAC,CAT H270H XP_006723263.1
XM_006723201.1 1226 Silent Mutation CAC,CAT H159H XP_006723264.1
XM_006723202.2 1226 Silent Mutation CAC,CAT H159H XP_006723265.1
XM_017026766.1 1226 Silent Mutation CAC,CAT H305H XP_016882255.1
XM_017026767.1 1226 Silent Mutation CAC,CAT H305H XP_016882256.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details