Product Details

SNP ID
rs181975111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55308056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAGCTGTTCAGCGTGTTCGAG[A/G]GCGACTCGACCACTGCGGCGGGAAC
Phenotype
MIM: 612720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX29 PubMed Links

Gene Details

Gene
DHX29
Gene Name
DEAH-box helicase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019030.2 309 Intron NP_061903.2
XM_006714653.2 309 Intron XP_006714716.1
XM_017009588.1 309 Intron XP_016865077.1
Gene
SKIV2L2
Gene Name
Ski2 like RNA helicase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015360.4 309 Missense Mutation AGC,GGC S15G NP_056175.3

View Full Product Details