Product Details

SNP ID
rs182050826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57326428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTACTCATCGGATCCTGACGC[C/T]GAGTCCTCTGAGCTGGGGGGAGTAC
Phenotype
MIM: 600843 MIM: 604328 MIM: 607104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RX3 PubMed Links

Gene Details

Gene
P2RX3
Gene Name
purinergic receptor P2X 3
There are no transcripts associated with this gene.

Gene
SSRP1
Gene Name
structure specific recognition protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003146.2 2593 Silent Mutation TCA,TCG S703S NP_003137.1
XM_017018180.1 2593 Silent Mutation TCA,TCG S837S XP_016873669.1
XM_017018181.1 2593 Intron XP_016873670.1
Gene
TNKS1BP1
Gene Name
tankyrase 1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details