Product Details
- SNP ID
-
rs181808385
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:159248378 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTATAGCATCACACATACCTGATG[C/T]GATGACCGGATCTTTCATAAGTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WDSUB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73004952] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WDSUB1
- Gene Name
- WD repeat, sterile alpha motif and U-box domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128212.2 |
1232 |
Missense Mutation |
ACA,GCA |
T423A |
NP_001121684.1 |
NM_001128213.1 |
1232 |
Missense Mutation |
ACA,GCA |
T423A |
NP_001121685.1 |
NM_001307994.1 |
1232 |
Missense Mutation |
ACA,GCA |
T331A |
NP_001294923.1 |
NM_152528.2 |
1232 |
Missense Mutation |
ACA,GCA |
T423A |
NP_689741.2 |
XM_005246355.2 |
1232 |
Intron |
|
|
XP_005246412.2 |
XM_006712341.3 |
1232 |
Missense Mutation |
ACA,GCA |
T254A |
XP_006712404.1 |
XM_011510753.2 |
1232 |
Missense Mutation |
ACA,GCA |
T423A |
XP_011509055.1 |
XM_011510754.2 |
1232 |
Missense Mutation |
ACA,GCA |
T336A |
XP_011509056.1 |
XM_017003481.1 |
1232 |
Missense Mutation |
ACA,GCA |
T423A |
XP_016858970.1 |
XM_017003482.1 |
1232 |
Missense Mutation |
ACA,GCA |
T300A |
XP_016858971.1 |
XM_017003483.1 |
1232 |
Missense Mutation |
ACA,GCA |
T185A |
XP_016858972.1 |
View Full Product Details