Product Details

SNP ID
rs182736053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38352378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGGCCCTGCCGCCGGGCTTGG[C/T]GCTCGAGCGGACCTGGGGCCCGGCG
Phenotype
MIM: 614515 MIM: 608788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR179 PubMed Links

Gene Details

Gene
GPR179
Gene Name
G protein-coupled receptor 179
There are no transcripts associated with this gene.

Gene
SOCS7
Gene Name
suppressor of cytokine signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014598.3 255 Missense Mutation GCG,GTG A45V NP_055413.1
XM_017024551.1 255 Missense Mutation GCG,GTG A109V XP_016880040.1
XM_017024552.1 255 Missense Mutation GCG,GTG A109V XP_016880041.1

View Full Product Details