Product Details

SNP ID
rs182850766
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:247965862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGCTTTCATACATACATTGTA[C/T]GTGTTTCAGCTTCCATTCTGTAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2AK2 PubMed Links

Gene Details

Gene
OR2AK2
Gene Name
olfactory receptor family 2 subfamily AK member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004491.1 531 Silent Mutation TAC,TAT Y177Y NP_001004491.1
Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 531 Intron NP_001291464.1
NM_175911.3 531 Intron NP_787107.1
XM_011544169.2 531 Intron XP_011542471.1

View Full Product Details