Product Details

SNP ID
rs182793699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196947345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCAGGGGTTACCACAAAGAGGC[G/T]GCAGAGCCATGGCCCACCAGCCACT
Phenotype
MIM: 605133 MIM: 611671
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NCBP2 PubMed Links

Gene Details

Gene
NCBP2
Gene Name
nuclear cap binding protein subunit 2
There are no transcripts associated with this gene.

Gene
NCBP2-AS2
Gene Name
NCBP2 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

Gene
PIGZ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025163.3 1737 Missense Mutation NP_079439.2
XM_011513190.1 1737 Missense Mutation XP_011511492.1
XM_011513192.2 1737 Missense Mutation XP_011511494.1
XM_017007240.1 1737 Missense Mutation XP_016862729.1
XM_017007241.1 1737 Missense Mutation XP_016862730.1
XM_017007242.1 1737 Missense Mutation XP_016862731.1
XM_017007243.1 1737 Missense Mutation XP_016862732.1

View Full Product Details