Product Details

SNP ID
rs182616636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35756934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGCGCTGGTCAAAGAGGCGTCC[A/G]GGCGCCGAAAGTCCGGGCAACGGGG
Phenotype
MIM: 610695 MIM: 607632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPB6 PubMed Links

Gene Details

Gene
HSPB6
Gene Name
heat shock protein family B (small) member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144617.2 96 Silent Mutation CCC,CCT P25P NP_653218.1
Gene
LIN37
Gene Name
lin-37 DREAM MuvB core complex component
There are no transcripts associated with this gene.

Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

Gene
PSENEN
Gene Name
presenilin enhancer gamma-secretase subunit
There are no transcripts associated with this gene.

View Full Product Details