Product Details

SNP ID
rs182629969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57602467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCTCCCACAATCTCACCAACA[C/T]TGGGCATCAACACGTCGACCTTTAA
Phenotype
MIM: 108410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NARS PubMed Links

Gene Details

Gene
NARS
Gene Name
asparaginyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004539.3 1631 Missense Mutation AAT,AGT N468S NP_004530.1
XM_005266700.1 1631 Missense Mutation AAT,AGT N467S XP_005266757.1

View Full Product Details