Product Details

SNP ID
rs182390255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31953791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATGGGCTGTTTTCGGGCTATG[C/T]TGACTTTGAGCTGTACAGACTCCAC
Phenotype
MIM: 613927 MIM: 138470 MIM: 154040 MIM: 600478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2 PubMed Links

Gene Details

Gene
C2
Gene Name
complement component 2
There are no transcripts associated with this gene.

Gene
CFB
Gene Name
complement factor B
There are no transcripts associated with this gene.

Gene
MIR1236
Gene Name
microRNA 1236
There are no transcripts associated with this gene.

Gene
NELFE
Gene Name
negative elongation factor complex member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002904.5 1187 Missense Mutation AAC,AGC N328S NP_002895.3
XM_006715205.3 1187 Missense Mutation AAC,AGC N323S XP_006715268.1
XM_011514913.2 1187 Missense Mutation AAC,AGC N273S XP_011513215.1
XM_017011299.1 1187 Missense Mutation AAC,AGC N268S XP_016866788.1
Gene
SKIV2L
Gene Name
Ski2 like RNA helicase
There are no transcripts associated with this gene.

View Full Product Details