Product Details
- SNP ID
-
rs182409100
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:105610533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCCACCCTCAGTAACTTACCTGT[C/T]GGCTGTGTCCTGGGCTGGATATACT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN7L1
PubMed Links
Gene Details
- Gene
- ATXN7L1
- Gene Name
- ataxin 7 like 1
View Full Product Details