Product Details

SNP ID
rs183376293
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:155321090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAATGAATAGAGAAAGGGTGCG[G/T]TGGTGGCTGAACGATGGAGACGAAT
Phenotype
MIM: 134629
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135821.1 676 Intron NP_001129293.1
NM_001135822.1 676 Intron NP_001129294.1
NM_001242824.1 676 Intron NP_001229753.1
NM_001242825.1 676 Intron NP_001229754.1
NM_002004.3 676 Intron NP_001995.1
XM_005244962.1 676 Intron XP_005245019.1
XM_005244963.1 676 Intron XP_005245020.1
Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 676 Intron NP_001098673.1
NM_001105204.1 676 Intron NP_001098674.1
NM_001105205.1 676 Intron NP_001098675.1
NM_001278227.1 676 Intron NP_001265156.1
NM_001278228.1 676 Intron NP_001265157.1
NM_001278229.1 676 Intron NP_001265158.1
NM_001278230.1 676 Intron NP_001265159.1
NM_014328.4 676 Intron NP_055143.2
XM_006711254.1 676 Intron XP_006711317.1
XM_006711256.1 676 Intron XP_006711319.1
XM_006711257.1 676 Intron XP_006711320.1
XM_017000891.1 676 Intron XP_016856380.1
XM_017000892.1 676 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 676 Silent Mutation NP_001034606.1

View Full Product Details