Product Details
- SNP ID
-
rs183429188
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130382136 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGACTTCCTGGACCTGCTGGTGG[C/G]GGAGAACCCGCACCTTGTCAGCAAG
- Phenotype
-
MIM: 114850
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPA1
PubMed Links
Gene Details
- Gene
- CPA1
- Gene Name
- carboxypeptidase A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001868.3 |
560 |
Missense Mutation |
GCG,GGG |
A137G |
NP_001859.1 |
View Full Product Details