Product Details

SNP ID
rs183029853
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:151838958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGCCGCAGGCTCCGCGGCCCCC[A/G]CCTTATAGCCAGCTTTTTCCAAGAG
Phenotype
MIM: 602943 MIM: 615653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD4D PubMed Links

Gene Details

Gene
C2CD4D
Gene Name
C2 calcium dependent domain containing 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136003.1 1081 Missense Mutation GCG,GTG A11V NP_001129475.1
XM_011509055.2 1081 Missense Mutation GCG,GTG A11V XP_011507357.1
XM_016999989.1 1081 Missense Mutation GCG,GTG A11V XP_016855478.1
Gene
LOC100132111
Gene Name
uncharacterized LOC100132111
There are no transcripts associated with this gene.

Gene
RORC
Gene Name
RAR related orphan receptor C
There are no transcripts associated with this gene.

Gene
THEM5
Gene Name
thioesterase superfamily member 5
There are no transcripts associated with this gene.

View Full Product Details