Product Details

SNP ID
rs182950647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19876505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTTGCTTCTTTCGCCACCCCA[A/G]AGATGATTACAGATTACCTAACAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K2 PubMed Links

Gene Details

Gene
OR4K2
Gene Name
olfactory receptor family 4 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005501.1 238 Missense Mutation AAG,GAG K80E NP_001005501.1

View Full Product Details