Product Details
- SNP ID
-
rs183541443
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:13105387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTCTCCTCCATAGCTTCGTCGGC[C/T]GCCTTGCCCCTGTGCGAGGGGAGGA
- Phenotype
-
MIM: 151440
MIM: 164005
MIM: 611669
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYL1
PubMed Links
Gene Details
- Gene
- LYL1
- Gene Name
- LYL1, basic helix-loop-helix family member
There are no transcripts associated with this gene.
- Gene
- NFIX
- Gene Name
- nuclear factor I X
There are no transcripts associated with this gene.
- Gene
- TRMT1
- Gene Name
- tRNA methyltransferase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136035.2 |
2271 |
Silent Mutation |
GCA,GCG |
A571A |
NP_001129507.1 |
NM_001142554.1 |
2271 |
Silent Mutation |
GCA,GCG |
A542A |
NP_001136026.1 |
NM_017722.3 |
2271 |
Silent Mutation |
GCA,GCG |
A571A |
NP_060192.1 |
XM_006722793.2 |
2271 |
Silent Mutation |
GCA,GCG |
A357A |
XP_006722856.1 |
XM_011528124.1 |
2271 |
Silent Mutation |
GCA,GCG |
A535A |
XP_011526426.1 |
XM_011528125.1 |
2271 |
Silent Mutation |
GCA,GCG |
A357A |
XP_011526427.1 |
XM_017026944.1 |
2271 |
Silent Mutation |
GCA,GCG |
A571A |
XP_016882433.1 |
XM_017026945.1 |
2271 |
Silent Mutation |
GCA,GCG |
A542A |
XP_016882434.1 |
XM_017026946.1 |
2271 |
Silent Mutation |
GCA,GCG |
A328A |
XP_016882435.1 |
XM_017026947.1 |
2271 |
Silent Mutation |
GCA,GCG |
A310A |
XP_016882436.1 |
View Full Product Details