Product Details

SNP ID
rs183959126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56576808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTACAAAAGGATTTTCCCCTA[A/G]TCATTTGTTGTATGGCAAGGATTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5M10 PubMed Links

Gene Details

Gene
OR5M10
Gene Name
olfactory receptor family 5 subfamily M member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004741.1 Intron NP_001004741.1

View Full Product Details