Product Details

SNP ID
rs183923514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121888887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGAGGAAGCGAGGCAGAGCGGA[A/G]GCTCCTCGCAGGCCGGCGTCGTGAC
Phenotype
MIM: 609695 MIM: 603146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPD PubMed Links

Gene Details

Gene
HPD
Gene Name
4-hydroxyphenylpyruvate dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171993.1 157 Intron NP_001165464.1
NM_002150.2 157 Intron NP_002141.1
Gene
PSMD9
Gene Name
proteasome 26S subunit, non-ATPase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261400.2 157 Missense Mutation AGC,GGC S11G NP_001248329.1
NM_002813.6 157 Missense Mutation AGC,GGC S11G NP_002804.2

View Full Product Details