Product Details

SNP ID
rs183681428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116252795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGGAACCTGAAGGATCTGGCG[C/T]TCTCCACTGGCTGCCTTGGTGGGCC
Phenotype
MIM: 605942
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSE PubMed Links

Gene Details

Gene
DSE
Gene Name
dermatan sulfate epimerase
There are no transcripts associated with this gene.

Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
There are no transcripts associated with this gene.

Gene
TSPYL4
Gene Name
TSPY like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021648.4 1304 Missense Mutation AAC,AGC N405S NP_067680.3

View Full Product Details