Product Details

SNP ID
rs183652831
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119110632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGACCCAGCAGTCCCCCGCTGC[C/T]GTCTCCATGGAGACCTACCACGTCA
Phenotype
MIM: 191350
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C2CD2L PubMed Links

Gene Details

Gene
C2CD2L
Gene Name
C2CD2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290474.1 1203 Silent Mutation GCC,GCT A174A NP_001277403.1
NM_014807.4 1203 Silent Mutation GCC,GCT A174A NP_055622.3
XM_006718948.3 1203 Silent Mutation GCC,GCT A174A XP_006719011.1
XM_011543076.2 1203 Silent Mutation GCC,GCT A25A XP_011541378.1
XM_017018625.1 1203 Silent Mutation GCC,GCT A174A XP_016874114.1
XM_017018626.1 1203 Silent Mutation GCC,GCT A174A XP_016874115.1
XM_017018627.1 1203 Silent Mutation GCC,GCT A174A XP_016874116.1
XM_017018628.1 1203 Silent Mutation GCC,GCT A25A XP_016874117.1
Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details