Product Details

SNP ID
rs183718167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31257752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTGTGGTCCTGGCTCAGACTG[C/T]CCCAGGTAAACAGAACCATGGAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB115 PubMed Links

Gene Details

Gene
DEFB115
Gene Name
defensin beta 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037730.1 89 Missense Mutation GCC,GTC A30V NP_001032819.1

View Full Product Details