Product Details

SNP ID
rs184434118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:69161432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCAGTTGCCACCTCCCCCTTCA[A/C]ATATTCGCTACAGCAGATGGGGCAG
Phenotype
MIM: 300420
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PJA1 PubMed Links

Gene Details

Gene
PJA1
Gene Name
praja ring finger ubiquitin ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032396.2 1622 Missense Mutation NP_001027568.1
NM_022368.4 1622 Missense Mutation NP_071763.2
NM_145119.3 1622 Missense Mutation NP_660095.1
XM_005262292.1 1622 Missense Mutation XP_005262349.1
XM_011531011.2 1622 Missense Mutation XP_011529313.1

View Full Product Details