Product Details

SNP ID
rs184452710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:47348895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGGCTACCTGCGGGTGGACAG[T/C]GTCCCGGGCCAGGAGGTGCTGAGCA
Phenotype
MIM: 180290
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RBP3 PubMed Links

Gene Details

Gene
RBP3
Gene Name
retinol binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002900.2 525 Missense Mutation NP_002891.1

View Full Product Details