Product Details

SNP ID
rs184171307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56015134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTTTCCTTTTCCACCAGATTC[A/G]GCTGTTGGCTGCGTCTCATCAAAAC
Phenotype
MIM: 172480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSPH PubMed Links

Gene Details

Gene
PSPH
Gene Name
phosphoserine phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004577.3 1188 Silent Mutation GCC,GCT A153A NP_004568.2
XM_005271773.1 1188 Silent Mutation GCC,GCT A153A XP_005271830.1
XM_005271774.1 1188 Silent Mutation GCC,GCT A153A XP_005271831.1
XM_005271775.1 1188 Silent Mutation GCC,GCT A153A XP_005271832.1
XM_005271776.1 1188 Silent Mutation GCC,GCT A153A XP_005271833.1
XM_006715760.1 1188 Silent Mutation GCC,GCT A153A XP_006715823.1
XM_006715761.1 1188 Silent Mutation GCC,GCT A108A XP_006715824.1
XM_011515461.1 1188 Silent Mutation GCC,GCT A153A XP_011513763.1
XM_017012466.1 1188 Silent Mutation GCC,GCT A153A XP_016867955.1
XM_017012467.1 1188 Silent Mutation GCC,GCT A153A XP_016867956.1
XM_017012468.1 1188 Silent Mutation GCC,GCT A108A XP_016867957.1
XM_017012469.1 1188 Silent Mutation GCC,GCT A108A XP_016867958.1

View Full Product Details