Product Details

SNP ID
rs184253479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54355974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCGAGGTGTCCGTCTCTCTGT[C/T]CTTCTCAGGAAGTCCATTGACTGTG
Phenotype
MIM: 602992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAIR1 PubMed Links

Gene Details

Gene
LAIR1
Gene Name
leukocyte associated immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289023.2 1007 Missense Mutation AAC,GAC N215D NP_001275952.2
NM_001289025.2 1007 Missense Mutation AAC,GAC N232D NP_001275954.2
NM_001289026.2 1007 Missense Mutation AAC,GAC N226D NP_001275955.2
NM_001289027.2 1007 Missense Mutation AAC,GAC N215D NP_001275956.2
NM_002287.5 1007 Missense Mutation AAC,GAC N233D NP_002278.2
NM_021706.4 1007 Missense Mutation AAC,GAC N216D NP_068352.2
XM_017026803.1 1007 Intron XP_016882292.1

View Full Product Details