Product Details

SNP ID
rs184626206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75235344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCCTGCTCGCAGGTCATCATA[G/T]AATCCCCCTGCGGCTGCTCCAGGGT
Phenotype
MIM: 193245
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COMTD1 PubMed Links

Gene Details

Gene
COMTD1
Gene Name
catechol-O-methyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144589.2 335 Missense Mutation TAT,TCT Y84S NP_653190.2
Gene
VDAC2
Gene Name
voltage dependent anion channel 2
There are no transcripts associated with this gene.

View Full Product Details