Product Details

SNP ID
rs185020773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101883701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCTCTTTTCCTTTGTCTGA[G/T]GCTCTTGTCTTTGTCTAAATCTGTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZMAT1 PubMed Links

Gene Details

Gene
ZMAT1
Gene Name
zinc finger matrin-type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011657.3 2093 Missense Mutation ATC,CTC I576L NP_001011657.2
NM_001282400.1 2093 Missense Mutation ATC,CTC I405L NP_001269329.1
NM_001282401.1 2093 Missense Mutation ATC,CTC I405L NP_001269330.1
XM_005262212.4 2093 Missense Mutation ATC,CTC I656L XP_005262269.2
XM_005262213.4 2093 Missense Mutation ATC,CTC I655L XP_005262270.2
XM_005262214.3 2093 Missense Mutation ATC,CTC I633L XP_005262271.2
XM_005262216.4 2093 Missense Mutation ATC,CTC I536L XP_005262273.2
XM_006724711.3 2093 Missense Mutation ATC,CTC I405L XP_006724774.1
XM_017029903.1 2093 Missense Mutation ATC,CTC I535L XP_016885392.1
XM_017029904.1 2093 Missense Mutation ATC,CTC I405L XP_016885393.1

View Full Product Details