Product Details

SNP ID
rs184720556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14956484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGCAAGGCCATACCTGGAAGA[C/T]GTGCCCATAGCGGTGATGAGGGCTT
Phenotype
MIM: 600637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC1A6 PubMed Links

Gene Details

Gene
SLC1A6
Gene Name
solute carrier family 1 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272087.1 1352 Intron NP_001259016.1
NM_001272088.1 1352 Intron NP_001259017.1
NM_005071.2 1352 Silent Mutation ACA,ACG T387T NP_005062.1
XM_006722842.1 1352 Silent Mutation ACA,ACG T387T XP_006722905.1
XM_006722844.1 1352 Silent Mutation ACA,ACG T387T XP_006722907.1
XM_017027152.1 1352 Silent Mutation ACA,ACG T387T XP_016882641.1

View Full Product Details