Product Details

SNP ID
rs184595978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231525275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTATAGAGCACGGGGTTGGCC[A/G]CCGAGCCCAGGTAGAAGAAGATGCC
Phenotype
MIM: 604153
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NMUR1 PubMed Links

Gene Details

Gene
NMUR1
Gene Name
neuromedin U receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006056.4 1147 Missense Mutation GCG,GTG A350V NP_006047.3
XM_006712195.3 1147 Intron XP_006712258.1
XM_006712196.3 1147 Intron XP_006712259.1
XM_011510487.2 1147 Missense Mutation GCG,GTG A327V XP_011508789.1
XM_011510488.2 1147 Intron XP_011508790.1
XM_011510489.2 1147 UTR 3 XP_011508791.1

View Full Product Details