Product Details

SNP ID
rs184575710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44775292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCAAAGTTTTTGTGCGAATTC[A/G]TCCTCCTGCAGAAAGATCTGGGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF15 PubMed Links

Gene Details

Gene
KIF15
Gene Name
kinesin family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020242.2 253 Missense Mutation CAT,CGT H34R NP_064627.1
XM_006713264.3 253 Intron XP_006713327.1
XM_011533964.2 253 Missense Mutation CAT,CGT H34R XP_011532266.1
XM_017006884.1 253 Missense Mutation CAT,CGT H34R XP_016862373.1
XM_017006885.1 253 Missense Mutation ATC,GTC I21V XP_016862374.1
XM_017006886.1 253 UTR 5 XP_016862375.1
XM_017006887.1 253 Missense Mutation CAT,CGT H34R XP_016862376.1
XM_017006888.1 253 Intron XP_016862377.1
XM_017006889.1 253 Missense Mutation CAT,CGT H34R XP_016862378.1
XM_017006890.1 253 Intron XP_016862379.1

View Full Product Details